Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1xr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030732.3 | NP_109657.2 | 9 | 1234 | ||
GenBank mRNA | AK029595.1 | - | 9 | 1033 | ||
AK033347.1 | - | 9 | 1104 | |||
AK036064.1 | - | 10 | 1345 | |||
AK136255.1 | - | 9 | 1056 |