Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTL6A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004301.4 | NP_004292.1 | 7 | 880 | ||
NM_177989.3 | NP_817126.1 | 7 | 736 | |||
NM_178042.3 | NP_829888.1 | 7 | 928 | |||
GenBank mRNA | AB015907.1 | - | 7 | 773 | ||
AB060168.1 | - | 7 | 705 | |||
AB061315.1 | - | 7 | 850 | |||
AF041474.1 | - | 7 | 803 | |||
AK021756.1 | - | 7 | 796 | |||
AK098691.1 | - | 7 | 834 | |||
AK223212.1 | - | 7 | 801 | |||
AL136608.1 | - | 7 | 820 | |||
BC000949.2 | - | 7 | 678 | 3445923 | ||
BC001391.2 | - | 7 | 721 | 3050818 | ||
BC036371.1 | - | 7 | 819 | 4827785 | ||
CR533529.1 | - | 7 | 667 |