Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028272.1 | - | 2 | 728 | ||
AK002290.1 | - | 2 | 622 | |||
AK135947.1 | - | 1 | 132 | |||
AK146657.1 | - | 2 | 690 | |||
AK158530.1 | - | 2 | 587 | |||
AK160924.1 | - | 2 | 624 | |||
AK164392.1 | - | 2 | 653 | |||
AK167718.1 | - | 2 | 627 | |||
AK168220.1 | - | 2 | 641 | |||
AK168291.1 | - | 2 | 641 | |||
BC012962.1 | - | 2 | 626 | 4188579 |