Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028272.1 | - | 3 | 986 | ||
AK002290.1 | - | 3 | 879 | |||
AK135947.1 | - | 2 | 391 | |||
AK146657.1 | - | 3 | 948 | |||
AK158530.1 | - | 3 | 845 | |||
AK160924.1 | - | 3 | 882 | |||
AK164392.1 | - | 3 | 911 | |||
AK167718.1 | - | 3 | 885 | |||
BC012962.1 | - | 3 | 884 | 4188579 |