Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB028272.1 | - | 1 | 339 | ||
AK002290.1 | - | 1 | 233 | |||
AK146657.1 | - | 1 | 301 | |||
AK160924.1 | - | 1 | 235 | |||
AK164392.1 | - | 1 | 264 | |||
AK167718.1 | - | 1 | 238 | |||
AK168220.1 | - | 1 | 252 | |||
AK168291.1 | - | 1 | 252 | |||
AK187532.1 | - | 1 | 42 | |||
AK200247.1 | - | 1 | 42 | |||
BC012962.1 | - | 1 | 237 | 4188579 | ||
BY251864.1 | - | 1 | 357 |