Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152922.3 | NP_690886.3 | 2 | 172 | ||
GenBank mRNA | AK148723.1 | - | 2 | 173 | ||
AY635583.1 | - | 2 | 173 | |||
BC055829.1 | - | 1 | 141 | |||
BC139484.1 | - | 2 | 141 | 8862506 | ||
BC141312.1 | - | 2 | 141 | 9055924 | ||
BC145632.1 | - | 2 | 141 | 9054200 |