Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030886.2 | NP_112148.2 | 33 | 7774 | ||
NM_198010.2 | NP_932127.2 | 32 | 7021 | |||
GenBank mRNA | AK031643.1 | - | 1 | 1540 | ||
AK043407.1 | - | 4 | 594 | |||
AK076402.1 | - | 5 | 2063 | |||
AK129193.1 | - | 9 | 3191 | |||
BC039213.1 | - | 5 | 1934 | |||
BC057195.1 | - | 5 | 1763 |