Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sorcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030889.2 | NP_112151.2 | 7, 8 | 1174 | ||
GenBank mRNA | AB093294.1 | - | 7, 8 | 1159 | ||
AK053853.1 | - | 7, 8 | 1144 | |||
AY004316.1 | - | 7, 8 | 1143 | |||
BC138873.1 | - | 7, 8 | 1100 | 8861895 | ||
BC138875.1 | - | 7, 8 | 1100 | 8861897 |