Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ift122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167763.1 | NP_001161235.1 | 17 | 2137 | ||
NM_031177.4 | NP_112454.2 | 17 | 2137 | |||
GenBank mRNA | AK031308.1 | - | 17 | 2025 | ||
AK044456.1 | - | 17 | 2137 | |||
AK077104.1 | - | 4 | 600 | |||
AK167330.1 | - | 17 | 2092 | |||
AK167412.1 | - | 17 | 2092 | |||
AK200860.1 | - | 1 | 26 | |||
BC038826.1 | - | 2 | 420 | |||
BC060621.1 | - | 17 | 2224 | |||
BC066083.1 | - | 17 | 2056 | 30355922 | ||
BC067415.1 | - | 17 | 2053 | 30063124 |