Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024281.2 | NP_077243.2 | 2 | 2099 | ||
NM_133626.2 | NP_598329.1 | 2 | 2099 | |||
GenBank mRNA | AF273683.1 | - | 2445 | |||
AF273684.1 | - | 2 | 2022 | |||
AF273685.1 | - | 3 | 1842 | |||
AF273686.1 | - | 3 | 1059 | |||
AF273687.1 | - | 3 | 1059 | |||
AF273688.1 | - | 4 | 1113 | |||
AF273689.1 | - | 3 | 909 | |||
AK019964.1 | - | 1 | 32 | |||
BC031452.1 | - | 2 | 2088 |