Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031183.2 | NP_112460.1 | 2 | 889 | ||
GenBank mRNA | AJ275988.1 | - | 1 | 118 | ||
AK137686.1 | - | 2 | 1112 | |||
AM295492.1 | - | 2 | 1133 | |||
AY338955.1 | - | 2 | 889 | |||
BC138818.1 | - | 1 | 835 | 8861840 | ||
BC138822.1 | - | 1 | 835 | 8861844 |