Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndst3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293682.1 | NP_001280611.1 | 6 | 824 | ||
NM_031186.3 | NP_112463.2 | 7 | 1937 | |||
GenBank mRNA | AF221095.1 | - | 7 | 1936 | ||
AK014990.1 | - | 5 | 977 | |||
AK015768.1 | - | 6 | 824 | |||
AK141945.1 | - | 8 | 2086 | |||
BC079622.1 | - | 7 | 1852 | 30535990 | ||
BC112404.1 | - | 7 | 1849 | 40060424 |