Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lamtor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031248.3 | NP_112538.1 | 3 | 336 | ||
GenBank mRNA | AJ277386.1 | - | 3 | 306 | ||
AK019006.1 | - | 3 | 331 | |||
BC031816.1 | - | 3 | 336 | 4189054 |