Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cstf2t | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031249.2 | NP_112539.2 | 1 | 1450 | ||
GenBank mRNA | AF322194.1 | - | 1 | 1295 | ||
AK030121.1 | - | 1 | 1289 | |||
AK034853.1 | - | 1 | 1288 | |||
AK050407.1 | - | 1 | 1406 | |||
AK082910.1 | - | 1 | 1322 | |||
AK084696.1 | - | 1 | 1284 | |||
AK173002.1 | - | 1 | 1260 | |||
BC026995.1 | - | 1 | 1258 | 5373155 |