Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031257.3 | NP_112547.1 | 5 | 495 | ||
GenBank mRNA | AF286161.1 | - | 5 | 449 | ||
AF418551.1 | - | 4 | 361 | |||
AK032325.1 | - | 5 | 505 | |||
AK042324.1 | - | 5 | 469 | |||
AK078212.1 | - | 5 | 474 | |||
AK088395.1 | - | 5 | 437 | |||
AK153117.1 | - | 5 | 467 | |||
BC010215.1 | - | 5 | 416 | 3498236 |