Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148930.3 | NP_683732.1 | 17 | 1549 | ||
GenBank mRNA | AJ309168.1 | - | 16 | 1384 | ||
AK040790.1 | - | 1 | 131 | |||
AK083714.1 | - | 1 | 37 | |||
AK134183.1 | - | 17 | 1544 | |||
AK146880.1 | - | 17 | 1544 | |||
BC003988.1 | - | 7 | 498 | |||
BC005719.1 | - | 12 | 1154 | |||
BC019766.1 | - | 17 | 1494 | |||
BC023854.1 | - | 17 | 1534 | 5346780 | ||
BC031899.1 | - | 17 | 1534 | 5362121 | ||
BC042419.1 | - | 15 | 4785 | |||
BC043058.1 | - | 17 | 1486 | 6419695 | ||
BC054729.1 | - | 17 | 1537 | 6419758 | ||
BC054820.1 | - | 15 | 4263 | |||
BC060985.1 | - | 16 | 1453 |