Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sacm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030692.4 | NP_109617.1 | 13 | 1042 | ||
GenBank mRNA | AJ245720.1 | - | 13 | 1172 | ||
AJ297526.1 | - | 13 | 1043 | |||
AK089174.1 | - | 13 | 1126 | |||
AK122390.1 | - | 13 | 1107 | |||
AK154759.1 | - | 13 | 1142 | |||
AK161111.1 | - | 13 | 1076 | |||
AK171080.1 | - | 13 | 1126 | |||
BC005489.1 | - | 4 | 251 | |||
BC117752.1 | - | 13 | 1043 | 40090556 | ||
BC117753.1 | - | 13 | 1043 | 40090558 |