Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024408.3 | NP_077719.2 | 24 | 4273 | ||
XM_005270901.3 | XP_005270958.1 | 25 | 6587 | |||
XM_011541519.2 | XP_011539821.1 | 24 | 9381 | |||
XM_011541520.2 | XP_011539822.1 | 24 | 6291 | |||
XM_017001372.1 | XP_016856861.1 | 24 | 3943 | |||
XM_017001373.1 | XP_016856862.1 | 24 | 3922 | |||
GenBank mRNA | AF308601.1 | - | 24 | 3988 | ||
AF315356.1 | - | 24 | 3988 |