Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tex11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167997.1 | NP_001161469.1 | 3 | 210 | ||
NM_031384.2 | NP_113561.2 | 3 | 210 | |||
GenBank mRNA | AF285572.1 | - | 3 | 208 | ||
AK029799.1 | - | 3 | 211 | |||
AK082794.1 | - | 3 | 202 | |||
BC116709.1 | - | 3 | 206 | 40096708 |