Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031388.2 | NP_113565.2 | 8 | 2061 | ||
GenBank mRNA | AF285570.1 | - | 5 | 1657 | ||
BC125454.1 | - | 3 | 1560 | 40129969 | ||
BC132154.1 | - | 3 | 1560 | 40130431 |