Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sytl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040085.2 | NP_001035174.1 | 15 | 2924 | ||
NM_001040087.2 | NP_001035176.1 | 6 | 1131 | |||
NM_001040088.2 | NP_001035177.1 | 6 | 1059 | |||
NM_001289583.1 | NP_001276512.1 | 16 | 2972 | |||
NM_001289584.1 | NP_001276513.1 | 16 | 2891 | |||
NM_001289585.1 | NP_001276514.1 | 7 | 1224 | |||
NM_001289586.1 | NP_001276515.1 | 7 | 1179 | |||
NM_031394.3 | NP_113571.2 | 15 | 2852 | |||
GenBank mRNA | AB050742.1 | - | 6 | 670 | ||
AB057754.1 | - | 15 | 2311 | |||
AB057755.1 | - | 9 | 2821 | |||
AB057756.1 | - | 7 | 1146 | |||
AB057757.1 | - | 2 | 1137 | |||
AB057760.1 | - | 15 | 2230 | |||
AB057761.1 | - | 14 | 2263 | |||
AB057762.1 | - | 14 | 2191 | |||
AB057763.1 | - | 3 | 1040 | |||
AI746961.1 | - | 6 | 604 | |||
AK027924.1 | - | 6 | 1042 | |||
AK051653.1 | - | 7 | 1225 | |||
AK173212.1 | - | 6 | 589 | |||
BC060665.1 | - | 7 | 2674 | |||
BC138714.1 | - | 14 | 2162 | 8861736 | ||
BC145398.1 | - | 15 | 2282 | 9053956 | ||
CA559030.1 | - | 5 | 565 |