Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030611.3 | NP_085114.1 | 8 | 942 | ||
GenBank mRNA | AK149458.1 | - | 8 | 943 | ||
BC056643.1 | - | 8 | 924 | 5098276 | ||
D45850.1 | - | 8 | 894 |