Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030611.3 | NP_085114.1 | 8, 9 | 971 | ||
GenBank mRNA | AK149458.1 | - | 8, 9 | 972 | ||
BC056643.1 | - | 8, 9 | 953 | 5098276 | ||
D45850.1 | - | 8, 9 | 923 |