Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tas1r3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031872.2 | NP_114078.1 | 2 | 313 | ||
GenBank mRNA | AB049994.1 | - | 2 | 326 | ||
AF311386.1 | - | 2 | 294 | |||
AF337039.1 | - | 2 | 313 | |||
AY032621.1 | - | 2 | 294 |
Set Membership: |
G-protein coupled receptor |