Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130187.1 | NP_001123659.1 | 5 | 846 | ||
NM_031878.2 | NP_114084.2 | 4 | 480 | |||
GenBank mRNA | AK159259.1 | - | 5 | 683 | ||
AK166123.1 | - | 5 | 639 | |||
BC005732.1 | - | 4 | 470 | 2647494 |