Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031842.2 | NP_114030.2 | 10 | 1180 | ||
GenBank mRNA | AK075611.1 | - | 6 | 517 | ||
AK208116.1 | - | 2 | 68 | |||
BC026783.1 | - | 10 | 1092 | 4481097 | ||
BC059921.1 | - | 10 | 1150 | |||
BC079839.1 | - | 10 | 1126 | 6810462 | ||
M25773.1 | - | 10 | 1136 | |||
U66620.1 | - | 10 | 1136 |