Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114386.1 | NP_001107858.1 | 14 | 1509 | ||
NM_031881.2 | NP_114087.2 | 13 | 1558 | |||
GenBank mRNA | AF277232.1 | - | 13 | 1191 | ||
AK004969.1 | - | 13 | 1251 | |||
AK031842.1 | - | 12 | 1179 | |||
AK042621.1 | - | 13 | 1132 | |||
AK163497.1 | - | 11 | 979 | |||
AK183000.1 | - | 3 | 199 | |||
BC039746.1 | - | 13 | 1202 | 3157137 | ||
BC071210.1 | - | 13 | 1179 | 4972426 |