Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031863.3 | NP_114069.1 | 9 | 901 | ||
GenBank mRNA | AK011998.1 | - | 9 | 887 | ||
AK012179.1 | - | 9 | 902 | |||
AK020040.1 | - | 9 | 906 | |||
AK076094.1 | - | 9 | 879 | |||
AK157569.1 | - | 9 | 926 | |||
BC018524.1 | - | 9 | 834 | 3588374 |