Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031863.3 | NP_114069.1 | 8 | 819 | ||
GenBank mRNA | AK011998.1 | - | 8 | 805 | ||
AK012179.1 | - | 8 | 820 | |||
AK020040.1 | - | 8 | 824 | |||
AK076094.1 | - | 8 | 797 | |||
AK157569.1 | - | 8 | 844 | |||
BC018524.1 | - | 8 | 752 | 3588374 |