Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031863.3 | NP_114069.1 | 4, 5 | 485 | ||
GenBank mRNA | AK011998.1 | - | 4, 5 | 471 | ||
AK012179.1 | - | 4, 5 | 486 | |||
AK020040.1 | - | 4, 5 | 487 | |||
AK020761.1 | - | 4, 5 | 981 | |||
AK076094.1 | - | 4, 5 | 463 | |||
AK088680.1 | - | 316 | ||||
AK157569.1 | - | 4, 5 | 510 | |||
BC018524.1 | - | 4, 5 | 418 | 3588374 |