Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002514.3 | NP_002505.1 | 5 | 1179 | ||
GenBank mRNA | AK294289.1 | - | 5 | 1006 | ||
AK312355.1 | - | 5 | 1045 | |||
AY082381.1 | - | 5 | 1030 | |||
BC015028.1 | - | 5 | 1025 | 3916150 | ||
CR457432.1 | - | 5 | 958 | |||
X96584.1 | - | 5 | 1030 |