Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032002.2 | NP_114391.1 | 3, 4 | 294 | ||
GenBank mRNA | AF083067.2 | - | 2, 3 | 172 | ||
AK080089.1 | - | 3, 4 | 272 |