Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACTN1 SIRNA Assays ›
Gene Symbol:
ACTN1
Gene Name:
actinin alpha 1
Gene Aliases:
BDPLT15
Chromosome Location:
Chr.14: 68874123 - 68979366 on Build GRCh38
UniGene ID:
Hs.235750
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s965
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ACTN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001102.3
NP_001093.1
6, 7
912
NM_001130004.1
NP_001123476.1
6, 7
912
NM_001130005.1
NP_001123477.1
6, 7
912
XM_011537265.2
XP_011535567.1
6, 7
704
XM_011537266.2
XP_011535568.1
6, 7
704
XM_011537267.2
XP_011535569.1
6, 7
704
XM_011537268.2
XP_011535570.1
6, 7
704
XM_017021720.1
XP_016877209.1
6, 7
704
XM_017021721.1
XP_016877210.1
6, 7
704
XM_017021722.1
XP_016877211.1
6, 7
849
XM_017021723.1
XP_016877212.1
6, 7
849
XM_017021725.1
XP_016877214.1
6, 7
1023
XM_017021726.1
XP_016877215.1
6, 7
1023
XM_017021727.1
XP_016877216.1
6, 7
849
XM_017021728.1
XP_016877217.1
6, 7
1023
GenBank mRNA
AK225782.1
-
6, 7
809
AK295099.1
-
6, 7
808
AK298496.1
-
6, 7
687
AK299362.1
-
6, 7
550
BC003576.1
-
6, 7
844
3547017
BC015766.1
-
6, 7
719
4856734
BT007207.1
-
6, 7
602
BX248020.1
-
6, 7
575
DQ496098.1
-
6, 7
785
EU716325.1
-
6, 7
622
FJ410030.1
-
6, 7
622
M95178.1
-
6, 7
713
X15804.1
-
6, 7
800
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
platelet degranulation
actin filament organization
platelet activation
platelet formation
platelet morphogenesis
regulation of apoptotic process
focal adhesion assembly
actin filament bundle assembly
negative regulation of cellular component movement
actin filament network formation
actin crosslink formation
platelet aggregation
regulation of nucleic acid-templated transcription
double-stranded RNA binding
integrin binding
calcium ion binding
protein binding
vinculin binding
ligand-dependent nuclear receptor transcription coactivator activity
protein homodimerization activity
ion channel binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-cp9cp:80/100.66.77.247:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline