Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168620.1 | NP_001162091.1 | 2 | 813 | ||
NM_032003.2 | NP_114392.1 | 2 | 899 | |||
GenBank mRNA | AF233377.1 | - | 2 | 876 | ||
AK032341.1 | - | 2 | 802 | |||
AK210356.1 | - | 1 | 309 | |||
AK218326.1 | - | 1 | 317 | |||
AK220510.1 | - | 2 | 842 | |||
BC089320.1 | - | 3 | 619 | |||
BC138977.1 | - | 2 | 814 | 8861999 | ||
BC138978.1 | - | 2 | 814 | 8862000 | ||
BC145484.1 | - | 2 | 814 |