Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168620.1 | NP_001162091.1 | 2 | 459 | ||
NM_032003.2 | NP_114392.1 | 2 | 545 | |||
GenBank mRNA | AF233377.1 | - | 2 | 522 | ||
AK032341.1 | - | 2 | 448 | |||
AK220510.1 | - | 2 | 488 | |||
BC089320.1 | - | 3 | 265 | |||
BC138977.1 | - | 2 | 460 | 8861999 | ||
BC138978.1 | - | 2 | 460 | 8862000 | ||
BC145484.1 | - | 2 | 460 |