Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032007.3 | NP_114396.3 | 9 | 1295 | ||
GenBank mRNA | AJ278461.1 | - | 9 | 1264 |