Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kremen1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032396.3 | NP_115772.2 | 9 | 1464 | ||
GenBank mRNA | AB059617.1 | - | 9 | 1427 | ||
AK141321.1 | - | 9 | 1476 | |||
BC049771.1 | - | 9 | 1388 | |||
BC082546.1 | - | 9 | 1464 | 6855459 | ||
BC138464.1 | - | 9 | 1472 | 8861484 | ||
BC138465.1 | - | 9 | 1472 | 8861485 |