Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252580.1 | NP_001239509.1 | 12 | 2049 | ||
NM_019729.3 | NP_062703.2 | 12 | 2016 | |||
GenBank mRNA | AB045709.1 | - | 11 | 1790 | ||
AF057146.1 | - | 11 | 1807 | |||
AK122195.1 | - | 12 | 1927 | |||
AK129003.1 | - | 12 | 2017 | |||
AK134058.1 | - | 12 | 1971 | |||
AK142881.1 | - | 9 | 1514 | |||
AK153015.1 | - | 11 | 1764 | |||
AK166540.1 | - | 10 | 1370 | |||
BC027052.1 | - | 2 | 569 | |||
BC050947.1 | - | 12 | 1981 | 6825981 | ||
BC061465.1 | - | 12 | 1981 | 6825981 | ||
BC066126.1 | - | 12 | 1979 | 30301665 |