Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032541.1 | NP_115930.1 | 2, 3 | 179 | ||
GenBank mRNA | AF297664.1 | - | 2, 3 | 179 | ||
AF503444.1 | - | 2, 3 | 179 | |||
BC021587.1 | - | 2, 3 | 162 | 5097807 |