Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam126a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053090.2 | NP_444320.2 | 9 | 1021 | ||
GenBank mRNA | AK155209.1 | - | 9 | 1022 | ||
AK161265.1 | - | 9 | 1215 | |||
BC056381.1 | - | 9 | 960 | 6852011 | ||
BC060692.1 | - | 9 | 1008 | 6410470 |