Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102.3 | NP_001093.1 | 16 | 2193 | ||
NM_001130004.1 | NP_001123476.1 | 16 | 2193 | |||
NM_001130005.1 | NP_001123477.1 | 16 | 2193 | |||
XM_011537265.2 | XP_011535567.1 | 16 | 2009 | |||
XM_011537266.2 | XP_011535568.1 | 16 | 2009 | |||
XM_011537267.2 | XP_011535569.1 | 16 | 1985 | |||
XM_011537268.2 | XP_011535570.1 | 16 | 1985 | |||
XM_017021720.1 | XP_016877209.1 | 16 | 2009 | |||
XM_017021721.1 | XP_016877210.1 | 16 | 1985 | |||
XM_017021722.1 | XP_016877211.1 | 16 | 2154 | |||
XM_017021723.1 | XP_016877212.1 | 16 | 2130 | |||
XM_017021725.1 | XP_016877214.1 | 16 | 2328 | |||
XM_017021726.1 | XP_016877215.1 | 16 | 2304 | |||
XM_017021727.1 | XP_016877216.1 | 16 | 2154 | |||
XM_017021728.1 | XP_016877217.1 | 16 | 2304 | |||
GenBank mRNA | AK098203.1 | - | 8 | 926 | ||
AK225782.1 | - | 16 | 2090 | |||
AK294317.1 | - | 10 | 1396 | |||
AK295099.1 | - | 16 | 2089 | |||
AK298496.1 | - | 16 | 1968 | |||
BC003576.1 | - | 16 | 2125 | 3547017 | ||
BC015766.1 | - | 16 | 2000 | 4856734 | ||
BC127124.1 | - | 1 | 70 | |||
BT007207.1 | - | 16 | 1883 | |||
BX641076.1 | - | 5 | 3333 | |||
DQ496098.1 | - | 16 | 2066 | |||
EU716325.1 | - | 16 | 1903 | |||
FJ410030.1 | - | 16 | 1903 | |||
M95178.1 | - | 16 | 1994 |