Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146087.2 | NP_666199.1 | 3 | 503 | ||
GenBank mRNA | AK076070.1 | - | 3 | 770 | ||
AK076264.1 | - | 3 | 764 | |||
AK078398.1 | - | 3 | 752 | |||
AK141421.1 | - | 3 | 757 | |||
AK146873.1 | - | 3 | 773 | |||
AK147165.1 | - | 3 | 748 | |||
AK152387.1 | - | 3 | 748 | |||
AK152793.1 | - | 3 | 768 | |||
AK166318.1 | - | 3 | 763 | |||
AK166429.1 | - | 3 | 745 | |||
AK167550.1 | - | 3 | 771 | |||
BC019740.1 | - | 3 | 503 | 3487913 | ||
BC025439.1 | - | 3 | 450 | 5038433 | ||
BC048081.1 | - | 3 | 536 | 6309504 |