Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053108.4 | NP_444338.2 | 2 | 332 | ||
GenBank mRNA | AB013137.1 | - | 2 | 298 | ||
AF109314.1 | - | 2 | 308 | |||
AF276917.1 | - | 2 | 257 | |||
AK013722.1 | - | 2 | 314 | |||
AK153086.1 | - | 2 | 312 | |||
BC012642.1 | - | 2 | 309 | 4206114 | ||
BC054418.1 | - | 2 | 294 | |||
BY272278.1 | - | 2 | 351 |