Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053068.3 | NP_444298.1 | 2, 3 | 302 | ||
GenBank mRNA | AF230805.1 | - | 2, 3 | 277 | ||
AK010812.1 | - | 2, 3 | 303 | |||
BC016593.1 | - | 2, 3 | 274 | |||
BC061043.1 | - | 2, 3 | 301 | 6773451 |