Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ice2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145618.3 | NP_663593.2 | 6 | 687 | ||
GenBank mRNA | AK134186.1 | - | 6 | 732 | ||
AK163917.1 | - | 6 | 615 | |||
AY244558.1 | - | 6 | 716 | |||
BC068183.1 | - | 6 | 687 | 30639735 |