Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhgc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033583.3 | NP_291061.1 | 1 | 1511 | ||
GenBank mRNA | AF464160.1 | - | 1 | 1407 | ||
AF464163.1 | - | 1 | 1511 | |||
AY013813.1 | - | 1 | 1382 | |||
BC138677.1 | - | 1 | 1455 | 8861698 | ||
BC138678.1 | - | 1 | 1569 | 8861699 |