Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000271.4 | NP_000262.2 | 15 | 2605 | ||
XM_005258277.1 | XP_005258334.1 | 15 | 2545 | |||
XM_005258278.4 | XP_005258335.1 | 15 | 2545 | |||
XM_005258279.2 | XP_005258336.1 | 15 | 2494 | |||
XM_006722479.3 | XP_006722542.1 | 15 | 2545 | |||
XM_011526015.1 | XP_011524317.1 | 15 | 2793 | |||
XM_017025784.1 | XP_016881273.1 | 15 | 2545 | |||
XM_017025785.1 | XP_016881274.1 | 15 | 2545 | |||
XM_017025786.1 | XP_016881275.1 | 15 | 2494 | |||
XM_017025787.1 | XP_016881276.1 | 15 | 2494 | |||
GenBank mRNA | AB209048.1 | - | 15 | 2366 | ||
AB971140.1 | - | 15 | 2331 | |||
AF002020.1 | - | 15 | 2454 | |||
AK293779.1 | - | 8 | 1409 | |||
AK295715.1 | - | 13 | 2245 | |||
BC063302.1 | - | 15 | 2428 | 30340517 |