Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhga6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033589.1 | NP_291067.1 | 1 | 1038 | ||
GenBank mRNA | AY013800.1 | - | 1 | 1038 | ||
BC059927.1 | - | 1 | 1173 | |||
BC080852.1 | - | 1 | 1083 | 6844625 |