Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhga11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033594.2 | NP_291072.1 | 1 | 543 | ||
GenBank mRNA | AY013794.1 | - | 1 | 375 | ||
BC059837.1 | - | 1 | 474 | 5719590 | ||
BC064675.1 | - | 1 | 543 | 6406939 |