Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhga12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033595.4 | NP_291073.1 | 1 | 625 | ||
GenBank mRNA | AY013795.1 | - | 1 | 88 | ||
BC054528.1 | - | 1 | 216 | 5719201 | ||
BC056998.1 | - | 1 | 625 | 6831906 | ||
BC066851.1 | - | 1 | 432 | 30086762 | ||
U88550.1 | - | 1 | 184 |